Learn More
Cyp2c7 Rabbit anti-Mouse, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech 55452-1-AP-20UL
 Lernen Sie weitere Varianten dieses
Artikelnummer. 16816925
  • 136.75 CHF / 20 Mikroliter
Verfügbar ab: 19-05-2025
Entdecken Sie weitere Sonderangebote
Beschreibung
The Cyp2c7 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of rat Cyp2c7. This antibody recognizes mouse, rat antigen. The Cyp2c7 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spezifikation
Spezifikation
Cyp2c7
Polyclonal
Unconjugated
20 μL
29298
IgG
Western Blot, Immunohistochemistry, ELISA
380 μg/mL
Rabbit
Primary
Mouse, Rat
Dokumentation
One moment while we fetch your results.
Name des Produkts
Cyp2c7 Rabbit anti-Mouse, Unconjugated, Polyclonal, Proteintech > 20 μL