Learn More
Cyclophilin A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
136.48 CHF - 385.92 CHF
Spezifikation
Antigen | Cyclophilin A |
---|---|
Konzentration | 0.32 mg/mL |
Anwendungen | Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence, Flow Cytometry |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | ||||||
16877283
|
Proteintech
10720-1-AP-20UL |
20 μL |
136.48 CHF
20 Mikroliter |
Lager vorrätig
Loggen Sie sich ein, um den Lagerbestand zu sehen |
Sie haben bereits ein Konto? Melden Sie sich an um den Preis und die Verfügbarkeit zu sehen. Neu hier? Registrieren Sie sich noch heute bei uns! |
|||||
16867283
|
Proteintech
10720-1-AP-150UL |
150 μL |
385.92 CHF
150 Mikroliter |
Lager vorrätig
Loggen Sie sich ein, um den Lagerbestand zu sehen |
Sie haben bereits ein Konto? Melden Sie sich an um den Preis und die Verfügbarkeit zu sehen. Neu hier? Registrieren Sie sich noch heute bei uns! |
|||||
Beschreibung
Immunophilins are a family of soluble cytosolic receptors capable of binding to one of two major immunosuppressant agents - cyclosporin A (CsA) or FK506. Proteins that bind FK506 are termed FK506 Binding Proteins (FKBPs) and those that bind cyclosporin A are called cyclophilins (CyP). Both CyP:CsA and FKBP:FK506 complexes have been shown to inhibit calcineurin, a calcium and calmodulin dependent protein phosphatase which has been implicated as an important signaling enzyme in T-cell activation, providing a possible mechanism of immunosuppression by CsA and FK506. Immunophilins function as peptidyl prolyl cis-trans-isomerases (PPIase) whose activity is inhibited by their respective immunosuppressant compounds. As PPIase′s, immunophilins accelerate folding of some proteins both in vivo and in vitro by catalyzing slow steps in the initial folding and rearrangement of proline containing proteins. Within the cyclophilin family, there are several different proteins which show a high degree of homology including CyPA, CyPB and CyPC. CyPA, also termed CyP-18, is the most abundant and ubiquitous cyclophilin found in all vertebrate tissues and is present in T-cells.Spezifikation
Cyclophilin A | |
Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence, Flow Cytometry | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
P10111, P17742, P62937 | |
25518, 268373, 5478 | |
Cyclophilin A Fusion Protein Ag1120 | |
Primary | |
-20°C | |
PPIA |
0.32 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Cyclophilin A, Cyclosporin A binding protein, CYPA, CYPH, PPIA, PPIase A, Rotamase A | |
PPIA | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.