Learn More
CoraLite™ Plus 488-conjugated FNTB Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech CL488-66783-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The FNTB antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FNTB. This antibody recognizes human, mouse antigen. The FNTB antibody has been validated for the following applications: IF/ICC, FC (Intra) analysis. This antibody is coralite™ plus 488.Spezifikation
FNTB | |
1F3B10 | |
CoraLite Plus 488 | |
Mouse | |
Protein G purification | |
Primary | |
Human, Mouse | |
44-45 kDa | |
Liquid |
Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
1000 μg/mL | |
FPTB; MGC31935 | |
Recombinant Protein | |
20 μL | |
2342 | |
Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
Conjugated Antibody | |
IgG1 |
Berichtigung von Produktinhalten
Name des Produkts
CoraLite™ Plus 488-conjugated FNTB Mouse Monoclonal Antibody, Proteintech > CoraLite™ Plus 488; 20 μL
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur