missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin 1 Rabbit anti-Canine, Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 13050-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Claudin-1 is a member of the transmembrane protein family claudins located in cell-cell tight junctions and it acts as a co-receptor for HCV entry into hepatic cells. Claudins are abundant in luminal epithelial sheets where they maintain epithelial cell polarity. Claudin-1 is expressed in most tissues such as bladder, fallopian tube, liver, pancreas, prostate, and skin.Spezifikation
Claudin 1 | |
Polyclonal | |
Unconjugated | |
CLDN1 | |
claudin 1, CLD1, CLDN1, ILVASC, SEMP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12737, 608207, 9076 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O88551, O95832 | |
CLDN1 | |
Claudin 1 Fusion Protein Ag3713 | |
20 μL | |
Primary | |
Human, Canine, Mouse | |
Antibody | |
IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Claudin 1 Rabbit anti-Canine, Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur