Learn More
Thermo Scientific™ CellSensor™ ISRE-bla HEK293T Cell Line
Beschreibung
Interferon cell signaling is a critical pathway involved in viral defense and autoimmune disease. The CellSensor™ ISRE-bla HEK 293T cell line is responsive to interferon alpha and can be used to probe the JAK/STAT signaling pathway. The CellSensor™ ISRE-bla HEK 293T cell line contains the β-lactamase reporter gene under the control of the interferon stimulated response element (ISRE). This cell line was engineered by transduction of the ISRE-bla construct into HEK 293T cells by lentivirus. Flow cytometry was then used to isolate a pool of clones responsive to interferon alpha a stimulation. A dose response curve for this assay against interferon alpha is available in the validation packet. Academic and non-profit customers, please inquire for special pricing.
Spezifikation
Spezifikation
| Assay-Eingabe | Zellbasiertes Beta-Lactamase-Reportergen |
| Zellenzustand | Teilende Zellen |
| Inhalt und Lagerung | CellSensor™ Zelllinien werden auf Trockeneis geliefert. Sofort nach Erhalt in Flüssigstickstoff lagern oder zur sofortigen Verwendung auftauen. |
| Zur Verwendung mit (Anwendung) | Zytokine, Signalweg |
| Produktlinie | CellSensor™ |
| Menge | 1 Fläschchen |
| Zieleintrag | Jak/STAT (Typ I IFN) Signalweg, TYK2, JAK1 |
| Typ | Teilende Zellen |
| System Type | CellSensor™ |
| Druggable Target | Signalweg |