missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD98 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15193-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This gene encodes subunit 1 of the splicing factor 3b protein complex. Splicing factor 3b, together with splicing factor 3a and a 12S RNA unit, forms the U2 small nuclear ribonucleoproteins complex. The splicing factor 3b/3a complex binds pre-mRNA upstream of the intron′s branch site in a sequence independent manner and may anchor the U2 snRNP to the pre-mRNA. Splicing factor 3b is also a component of the minor U12-type spliceosome. The carboxy-terminal two-thirds of subunit 1 have 22 non-identical, tandem HEAT repeats that form rod-like, helical structures. Alternative splicing results in multiple transcript variants encoding different isoforms.Spezifikation
CD98 | |
Polyclonal | |
Unconjugated | |
SLC3A2 | |
4F2, 4F2 heavy chain antigen, 4F2HC, 4T2HC, CD98, CD98HC, MDU1, NACAE, SLC3A2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6520 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry, Western Blot, Immunocytochemistry | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P08195 | |
SLC3A2 | |
CD98/SLC3A2 Fusion Protein Ag7280 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
CD98 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur