missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45 Mouse, PE-Cy7, Clone: 2D1, enQuireBio™
Mouse Monoclonal Antibody
Spezifikation
Antigen | CD45 |
---|---|
Klon | 2D1 |
Konzentration | 5μL/Test |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Spezifikation
CD45 | |
5μL/Test | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
5788 | |
IgG1 κ | |
Protein A/G Purified |
2D1 | |
Flow Cytometry | |
PE-Cyanine7 | |
RUO | |
NP_002829 | |
PTPRC | |
Primary | |
Store at 2-4°C. |