missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD38 Antibody (HIT2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Bio-Techne NB500-510
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CD38 Monoclonal specifically detects CD38 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.
Spezifikation
CD38 | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 2 μg/mL, Flow Cytometry 1 to 4 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 10 μg/mL, Immunohistochemistry-Frozen 1:200, CyTOF-ready | |
CD38 | |
Human thymocytes in foetus | |
Affinity Purified | |
RUO | |
Primary | |
The antibody HIT2 reacts with CD38 (T10), a 45 kDa type II transmembrane glycoprotein strongly expressed mainly on plasma cells and activated T and B lymphocytes; it is an antigenic marker of lymphoid cells. | |
Store at 4C. Do not freeze. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
HIT2 | |
Unconjugated | |
ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, cADPr hydrolase 1, CD38 antigen, CD38 antigen (p45), CD38 molecule, Cyclic ADP-ribose hydrolase 1, EC 3.2.2.5, NAD(+) nucleosidase, T10 | |
Mouse | |
45 kDa | |
0.1 mg | |
Apoptosis, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Lipid and Metabolism, Neuroscience, Stem Cell Markers | |
952 | |
Human | |
IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
CD38 Antibody (HIT2) - BSA Free, Novus Biologicals™ > 0.1mg; Unlabeled
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur