missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD206 Mouse anti-Human, Mouse, Clone: 2A6A10, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 60143-1-IG-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This antibody recognizes the intracellular carboxy-terminal part of CD206 and MRC1L1.
CD206 (macrophage mannose receptor, MMR), also known as mannose receptor C1 (MRC1), is a type I transmembrane glycoprotein serving as pattern recognition receptor for carbohydrate groups on the surface of bacteria, fungi and other pathogens. Expressed mainly on tissue macrophages and dendritic cells, CD206 mediates endocytosis of these pathogens and presentation of their antigens to the adaptive immune system. CD206 can also be detected in a soluble form in human plasma and is elevated in patients with acute sepsis.Spezifikation
CD206 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P22897, Q61830 | |
Mrc1 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG2a |
Immunoprecipitation, Western Blot, Flow Cytometry | |
2A6A10 | |
Unconjugated | |
MRC1 | |
CD206, CLEC13D, Macrophage mannose receptor 1, Mannose receptor, mannose receptor, C type 1, MMR, MRC1 | |
Mouse | |
Caprylic Acid Precipitation | |
RUO | |
17533, 4360 | |
-20°C | |
Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
CD206 Mouse anti-Human, Mouse, Clone: 2A6A10, Proteintech > 20 μL; Unconjugated
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur