Learn More
CCL19/MIP-3 beta Antibody (RM0171-10J38), Alexa Fluor™ 532, Novus Biologicals™
Rat Monoclonal Antibody
Marke:  Bio-Techne NBP2-12098AF532
Artikelnummer. 18125060
  • 693.00 CHF / 0.10 Milliliter
Verfügbar ab: 30-06-2025
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
CCL19/MIP-3 beta Monoclonal specifically detects CCL19/MIP-3 beta in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
CCL19/MIP-3 beta
Monoclonal
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
CCL19
Mouse recombinant CCL19/MIP-3 beta
0.1 mL
6363
Store at 4°C in the dark.
Western Blot
RM0171-10J38
Western Blot
beta chemokine exodus-3, Beta-chemokine exodus-3, CC chemokine ligand 19, C-C motif chemokine 19, chemokine (C-C motif) ligand 19, CKb11, EBI1-ligand chemokine, ELCMIP-3-beta, Epstein-Barr virus-induced molecule 1 ligand chemokine, exodus-3, Macrophage inflammatory protein 3 beta, macrophage inflammatory protein 3-beta, MGC34433, MIP-3b, MIP3BCK beta-11, SCYA19EBI1 ligand chemokine, small inducible cytokine subfamily A (Cys-Cys), member 19, Small-inducible cytokine A19
Rat
Protein A or G purified
Primary
Mouse
IgG2
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen
Berichtigung von Produktinhalten
Name des Produkts
CCL19/MIP-3 beta Antibody (RM0171-10J38), Alexa Fluor™ 532, Novus Biologicals™ > CCL19/MIP-3 beta Antibody; Alexa Fluor™ 532; 0.1 mL