Learn More
Novus Biologicals™ CARS2 Recombinant Protein Antigen
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Marke: Novus Biologicals™ NBP2-32597PEP
Beschreibung
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human CARS2. The CARS2 Recombinant Protein Antigen is derived from E. coli. The CARS2 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.This is a blocking peptide for NBP2-32597. This is a human recombinant protein expressed in E. coli with a N-terminal His-ABP tag and purified with IMAC chromatography.

Spezifikation
79587 | |
Chromatography | |
0.5mg/mL | |
PBS and 1M Urea, pH 7.4. | |
CARS2 | |
30kDa | |
0.1 mL | |
E.Coli | |
VFGAIISYFEQFFETVGISLANQQYVSGDGSEATLHGVVDELVRFRQKVRQFALAMPEATGDARRQQLLERQPLLEACDTLRRGLTAHGINIKDRSSTTSTWELLDQR |
Human | |
>80% | |
Store at -20°C. Avoid freeze-thaw cycles. | |
Blocking/Neutralizing, Control | |
Unmarkiert | |
CARS2 | |
RUO | |
This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-32597. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Nur für Forschungszwecke