missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calreticulin Antibody (1G6A7), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Bio-Techne NBP1-47518AF594
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Calreticulin Monoclonal specifically detects Calreticulin in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).
Spezifikation
Calreticulin | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
CALR | |
Calreticulin Antibody (1G6A7) was developed against a synthetic peptide corresponding to the C-terminus (EEEDVPGQAKDELC) of human Calreticulin, conjugated to KLH. [UniProt# P27797] | |
Autophagy, Cancer, Cellular Markers, ER Markers, Immunology, Membrane Trafficking and Chaperones, Neurodegeneration, Neuroscience, Signal Transduction | |
811 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1G6A7 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular) | |
autoantigen Ro, calregulin, calreticulin, cC1qR, CRP55, CRT, CRTC, Endoplasmic reticulum resident protein 60, ERp60, FLJ26680, grp60, RO, Sicca syndrome antigen A (autoantigen Ro; calreticulin), SSAHACBP | |
Mouse | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG2a |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Calreticulin Antibody (1G6A7), Alexa Fluor™ 594, Novus Biologicals™ > Calreticulin Antibody; Alexa Fluor™ 594; 0.1 mL
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur