missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calponin-1 Mouse, Clone: rCNN1/832, enQuireBio™
Mouse Monoclonal Antibody
Marke: EnquireBio 1264-MSM6-P0
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
Calponin-1 | |
Cytoplasmic | |
rCNN1/832 | |
Unconjugated | |
XP_005259798 | |
Mouse | |
34kDa | |
Protein A/G Purified | |
RUO | |
1264 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
CNN1 | |
Recombinant full-length human CNN1 protein | |
Myoepithelial cells in breast ducts | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |