missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Unity Lab Services™ Calibration Certificate Cofrac Temperature Three Fixed Points, -20°C, 0°C, 120°C
ULS offers a full range of support services and programs for all instruments and supplies, regardless of manufacturer. Maximize your technology investments by anticipating and avoiding problems before they occur.
Marke: Unity Lab Services™ UMV-P935B
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- Calibration by comparison with a reference thermometer in baths
- Type of device: temperature measuring thermometric chain or expansion thermometer, at fixed values of -20°C, 0°C, 120°C
- Our best uncertainties for a thermometer with a resolution of 0.01°C (k = 2) +/- 0.02°C at 0°C; +/- 0.05°C to -20°C;+/- 0.08°C to 120°C
- Our best uncertainties for a thermometer with a resolution of 0.1°C (k = 2) +/- 0.1°C to 0°C; +/- 0.2°C to -20°C and 120°C
- COFRAC Accreditation Number 2-5984
- Calibration, scope available on www.cofrac.fr
- Average time: 1 week
Spezifikation
| Thermometers and Temperature Recorders | |
| Cofrac | |
| Kalibrierungszertifikat |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Unity Lab Services™ Calibration Certificate Cofrac Temperature Three Fixed Points, -20°C, 0°C, 120°C >
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur