missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Met Monoclonal Antibody (85503), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA523760
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
In sandwich ELISAs, no cross-reactivity with recombinant human (rh) EGFR or rhIGF-1 R is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
c-Met Monoclonal antibody specifically detects c-Met in Human samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry.
Spezifikation
c-Met | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P08581 | |
MET | |
Mouse myeloma cell line NS0-derived recombinant human HGF R/c-MET Glu25-Thr932. | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunohistochemistry, Immunocytochemistry | |
85503 | |
Unconjugated | |
MET | |
AI838057; AUTS9; bHLHe59; CG1705; CG1705-PA; CG1705-PB; cmet; c-met; c-met proto-oncogene; c-Met receptor tyrosine kinase; c-Met/HGF receptor; D249; DFNB97; Dmel\CG1705; Dmel_CG1705; DmMet; EC 2.7.10.1; Hepatocyte growth factor receptor; HGF; HGF receptor; HGF receptor c-Met; HGF/SF receptor; HGFR; HGF-SF receptor; juvenile hormone resistance; met; met proto-oncogene; met proto-oncogene (hepatocyte growth factor receptor); met proto-oncogene tyrosine kinase; MET proto-oncogene, receptor tyrosine kinase; Met/Met1; Met; protein tyrosin kinase; methoprene tolerant; Methoprene-tolerant; Methoprene-tolerant protein; Met-PA; Met-PB; Mett; oncogene MET; Par4; Proto-oncogene c-Met; RCCP2; resistance (1) juvenile H; resistance to juvenile hormone; Rst(1)JH; sc MET; scatter factor receptor; SF receptor; soluble c met; tyrosine-protein kinase Met | |
Mouse | |
Protein A/G | |
RUO | |
4233 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
c-Met Monoclonal Antibody (85503), Invitrogen™ > 500 μg; Unconjugated
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur