Learn More
Robu™ Büchner Filter Funnels with Porosity 3 VitraPOR™ Glass Filter Discs
Robu™ Büchner Filter Funnels with Glass Filter Discs are made of pure borosilicate 3.3 glass and are highly resistant to most chemicals, including solvents, acids, and alkalis. The VitraPOR™ filter disc is rated as Porosity 3 (Pore size: 16 to 40 μm, Standard: ISO 4793-80).
Marke: Robu™ 21503
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- Product Type: Büchner Filter Funnel
- Material: Borosilicate 3.3 Glass (Body), VitraPOR™ Glass (Filter)
- Porosity: Grade 3
- Pore Size: 16-40 μm
- Standard: ISO 4793-80
- For Use With (Application): Filtration
- For Use With (Equipment): Guko Rubber Gaskets (e.g. 10430652)
Spezifikation
40 mm (OD); 36 mm (ID) | |
10 mm | |
53 mm | |
Äußerst widerstandsfähig gegen die meisten Chemikalien wie Lösungsmittel, Säuren und Alkalien | |
Guko Rubber Gaskets (e.g. 10430652) | |
16 to 40 μm | |
Büchner-Trichter |
50 ml | |
Borosilikatglas | |
ISO 4793-80 | |
40 mm | |
Grade 3 | |
1.7 oz. |