missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BLK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 10510-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
BLK is a protein kinases, which are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The STE group (homologs of yeast Sterile 7, 11, 20 kinases) consists of 50 kinases related to the mitogen-activated protein kinase (MAPK) cascade families (Ste7/MAP2K, Ste11/MAP3K, and Ste20/MAP4K). MAP kinase cascades, consisting of a MAPK and one or more upstream regulatory kinases (MAPKKs) have been best characterized in the yeast pheromone response pathway. Pheromones bind to Ste cell surface receptors and activate yeast MAPK pathway.Spezifikation
BLK | |
Polyclonal | |
Unconjugated | |
BLK | |
B lymphocyte kinase, B lymphoid tyrosine kinase, BLK, p55 Blk, Tyrosine protein kinase Blk | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12143, 364403, 640 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P16277, P51451 | |
Blk | |
BLK Fusion Protein Ag0795 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
BLK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur