missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-6 Mouse, Clone: BCL6/1475, enQuireBio™
Mouse Monoclonal Antibody
Marke: EnquireBio 604-MSM5-P0
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
Bcl-6 | |
Nuclear | |
BCL6/1475 | |
Unconjugated | |
NP_001124317 | |
Mouse | |
95kDa | |
Protein A/G Purified | |
RUO | |
604 | |
IgG1 κ |
Western Blot | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
BCL6 | |
Recombinant human bcl-6 protein fragment (aa256-389) | |
Raji or Ramos cells. Tonsil | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |