Learn More
PEBP1, Human, Matched Antibody Pair (AP46), Abnova™
To detect and quantify protein level of human
Marke:  Abnova H00005037-AP46.1Set
Artikelnummer. 16112404
  • 872.00 CHF / 1 Set
Nur noch null auf Lager
Verfügbar ab: 22-07-2025
Zum Warenkorb hinzufügen
Includes: Capture antibody: mouse monoclonal anti-PEBP1, IgG Mix Kappa (100μg) , Detection antibody: biotinylated mouse monoclonal anti-PEBP1, IgG Mix Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Spezifikation
PEBP1
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human PEBP1.
HCNP/PBP/PEBP/RKIP
Mouse
RUO
5037
Human
Liquid
ELISA
Biotin
PEBP1
PEBP1
1 Set
Primary
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-PEBP1, IgG Mix Kappa (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-PEBP1, IgG Mix Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
One moment while we fetch your results.
Zertifikate
Aktionen
Aktionen
Berichtigung von Produktinhalten
Name des Produkts
PEBP1, Human, Matched Antibody Pair (AP46), Abnova™ > 1 Set