missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL1B, Human, Matched Antibody Pair (AP71), Abnova™
To detect and quantify protein level of human
Marke: Abnova H00003553-AP71.1Set
Includes: Capture antibody: rabbit purified polyclonal anti-IL1B (125μg) , Detection antibody: mouse monoclonal anti-IL1B, IgG1 Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Spezifikation
IL1B | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human IL1B. | |
IL-1/IL1-BETA/IL1F2 | |
Mouse, Rabbit | |
RUO | |
3553 | |
Human | |
Liquid |
ELISA | |
Unconjugated | |
IL1B | |
IL1B | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit purified polyclonal anti-IL1B (125 ug) 2. Detection antibody: mouse monoclonal anti-IL1B, IgG1 Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
IL1B, Human, Matched Antibody Pair (AP71), Abnova™ > 1 Set
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur