Learn More
IL13, Human, Matched Antibody Pair (AP41), Abnova™
To detect and quantify protein level of human
Marke: Abnova H00003596-AP41.1Set
Includes: Capture antibody: mouse monoclonal anti-IL13 (100μg) , Detection antibody: biotinylated mouse monoclonal anti-IL13, IgG2a Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Spezifikation
IL13 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human IL13. | |
ALRH/BHR1/IL-13/MGC116786/MGC116788/MGC116789/P600 | |
Mouse | |
RUO | |
3596 | |
Human | |
Liquid |
ELISA | |
Biotin | |
IL13 | |
IL13 | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-IL13 (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-IL13, IgG2a Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Berichtigung von Produktinhalten
Name des Produkts
IL13, Human, Matched Antibody Pair (AP41), Abnova™ > 1 Set