Learn More
anti-EPHB3, Human, Matched Antibody Pair, Abnova™
To detect and quantify protein level of human
Marke: Abnova H00002049-AP41.1Set
Includes: Capture antibody: mouse monoclonal anti-EPHB3, IgG1 Kappa (100μg) , Detection antibody: biotinylated mouse monoclonal anti-EPHB3, IgG2a Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Spezifikation
EPHB3 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human EPHB3. | |
ETK2/HEK2/TYRO6 | |
Mouse | |
RUO | |
2049 | |
Human | |
Liquid |
ELISA | |
Biotin | |
EPHB3 | |
EPHB3 | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-EPHB3, IgG1 Kappa (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-EPHB3, IgG2a Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Berichtigung von Produktinhalten
Name des Produkts
anti-EPHB3, Human, Matched Antibody Pair, Abnova™ > 1 Set