Learn More
anti-DLX1, Human, Matched Antibody Pair, Abnova™
To detect and quantify protein level of human
Marke:  Abnova H00001745-AP61.1Set
Artikelnummer. 16121964
  • 872.00 CHF / 1 Set
Nur noch null auf Lager
Verfügbar ab: 25-07-2025
Zum Warenkorb hinzufügen
Includes: Capture antibody: mouse monoclonal anti-DLX1 (100μg) , Detection antibody: rabbit MaxPab affinity purified polyclonal anti-DLX1 (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Spezifikation
DLX1
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human DLX1.
DLX1
1 Set
Primary
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-DLX1 (100 μg) 2. Detection antibody: rabbit MaxPab™ affinity purified polyclonal anti-DLX1 (50 μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
ELISA
Unconjugated
DLX1
Mouse, Rabbit
RUO
1745
Human
Liquid
Dokumentation
One moment while we fetch your results.
Zertifikate
Aktionen
Aktionen
Berichtigung von Produktinhalten
Name des Produkts
anti-DLX1, Human, Matched Antibody Pair, Abnova™ > 1 Set