missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-BMPR1B, Human, Matched Antibody Pair, Abnova™
To detect and quantify protein level of human
Marke: Abnova H00000658-AP41.1Set
Includes: Capture antibody: mouse monoclonal anti-BMPR1B, IgG1 Kappa (100μg) , Detection antibody: biotinylated mouse monoclonal anti-BMPR1B, IgG1 Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Spezifikation
BMPR1B | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human BMPR1B. | |
ALK-6/ALK6/CDw293 | |
Mouse | |
RUO | |
658 | |
Human | |
Liquid |
ELISA | |
Biotin | |
BMPR1B | |
BMPR1B | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-BMPR1B, IgG1 Kappa (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-BMPR1B, IgG1 Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
anti-BMPR1B, Human, Matched Antibody Pair, Abnova™ > 1 Set
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur