Learn More
Invitrogen™ Aldosterone Competitive ELISA Kit
ELISA
722.00 CHF - 6220.00 CHF
Spezifikation
Assay-Bereich | 3.906 bis 4000 pg/ml |
---|---|
Assay-Empfindlichkeit: | 4,97 pg/mL |
Beschreibung | Aldosterone Competitive ELISA Kit |
Konjugat | HRP |
Produkttyp | ELISA-Kit |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | ||||||
15835853
|
Invitrogen™
EIAALD |
96 Tests |
722.00 CHF
96 Stück |
Verfügbar ab: 22-07-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
Sie haben bereits ein Konto? Melden Sie sich an um den Preis und die Verfügbarkeit zu sehen. Neu hier? Registrieren Sie sich noch heute bei uns! |
|||||
17855177
|
Invitrogen™
EIAALDX10 |
10 x 96 Tests |
6220.00 CHF
10 Stück |
Verfügbar ab: 28-07-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
Sie haben bereits ein Konto? Melden Sie sich an um den Preis und die Verfügbarkeit zu sehen. Neu hier? Registrieren Sie sich noch heute bei uns! |
|||||
Beschreibung
The Aldosterone Competitive ELISA quantitates aldosterone in extracted serum,plasma,urine,dried fecal extracts or cell culture medium.
Principle of the method
The Aldosterone Competitive ELISA Kit research-use-only kit is designed to quantitatively measure aldosterone present in extracted serum and plasma,or in urine,extracted dried fecal samples,and tissue culture media samples. An aldosterone stock solution is provided to generate a standard curve for the assay and all samples should be read off the standard curve. Standards or diluted samples are pipetted into the provided plate coated with a capture antibody. An aldosterone-peroxidase conjugate is added to the standards and samples in the wells. The binding reaction is initiated by the addition of a polyclonal antibody to aldosterone to each well. After incubation,the plate is washed and substrate is added. The substrate reacts with the bound aldosterone-peroxidase conjugate. After a short incubation,the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader capable of measuring at 450 nm.
Rigorous validation
Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity,specificity,precision,and lot-to-lot consistency. See manual for more information on validation.
Aldosterone is a hormone produced by the adrenal glands that helps regulate blood pressure through increasing both the release of potassium and the uptake of water and sodium by the kidneys. It is essential for sodium conservation in the kidney, salivary glands, sweat glands and colon. It plays a central role in the regulation of the plasma sodium (Na+), the extracellular potassium (K+) and arterial blood pressure. It does so mainly by acting on the mineralocorticoid receptors in the distal tubules and collecting ducts of the nephron. It influences the reabsorption of sodium and excretion of potassium (from and into the tubular fluids, respectively) of the kidney, thereby indirectly influencing water retention or loss, blood pressure and blood volume. When dysregulated, aldosterone is pathogenic and contributes to the development and progression of cardiovascular and renal disease. Aldosterone has exactly the opposite function of the atrial natriuretic hormone secreted by the heart.Spezifikation
3.906 bis 4000 pg/ml | |
Aldosterone Competitive ELISA Kit | |
ELISA-Kit | |
ELISA, Protein-Assays, Aktivitäts-Assays, Proteinbiologie, Reproduktive Biologie, Entzündungen, Oxidativer Stress, Metabolismus, Zellsignalweiterleitung, Epigenetik, Hämatologie, DNA-Schaden, Krebsbiologie, Immunologie, Neurowissenschaften, Neurobiologie, Steroid-Hormon-Nachweis | |
19.5 % | |
Vorbeschichtete 96-Well-Platte, Standard, HRP-konjugierter Nachweisantikörper., HRP-konjugierter Nachweisantikörper., Waschpuffer, Chromogen, Stopplösung, Dichtungsfolien für Mikrotiterplatten | |
RUO | |
Stuhlextrakt, 100 μl; Plasma, 100 μl; Serum, 100 μl; Überstand, 100 μl; Urin, 25 μl | |
Chemisch | |
16 Std 45 Min |
4,97 pg/mL | |
HRP | |
Stuhlextrakt, Plasma, Serum, Überstand, Urin | |
Kolorimetrisches Mikrotiterplatten-Lesegerät | |
6.9 % | |
HRP | |
Reproduktive Biologie | |
-20 °C | |
1 h 20 min |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.