Learn More
Melford Agarose Low Melt
Ideal gelling properties for use in cloning assays or for recovery of heat labile samples following electrophoresis.
Marke: Melford A20070-100.0
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Ideal gelling properties for use in cloning assays or for recovery of heat labile samples following electrophoresis.
Ideal gelling properties for use in cloning assays or for recovery of heat labile samples following electrophoresis.
Gel Strength, 1%: >200 g/cm2
Gelling Temperature: 26°C to 30°C
Melting Temperature: <65°C
Sulfate: <0.10%
EEO: <0.10
DNase and RNase: None Detected
Spezifikation
Agarose, Low Melt | |
9012-36-6 | |
100 g | |
Molecular Biology |