Learn More
Beschreibung
Activated carbon, also called activated charcoal, is a form of carbon processed to have small, low-volume pores that increase the surface area available for adsorption or chemical reactions. Granular activated carbon (GAC) has a relatively larger particle size (and, therefore, smaller external surface area) compared to powdered activated carbon. GAC is suitable for adsorption of gases and vapors, because they diffuse rapidly. Granulated carbons are used for water treatment, deodorization and separation of components of flow system and is also used in rapid mix basins.
- Non Solubles in Nitric Acid: <5%
- Non Solubles in Water: <2%
- Heavy Metals as Lead (Pb): <0.01%
- Iron (Fe): <0.6%
- Zinc (Zn): <0.15%
- Residue after Ignition: <2% at 600°C
Spezifikation
Spezifikation
Chemischer Name oder Material | Carbon |
Physikalische Form | Fest |
Schmelzpunkt | 3550°C |
Dichte | 1.8 to 2.1 g/cm3 |
Farbe | Black |
Menge | 500 g |
CAS | 7440-44-0 |
Partikelgröße | ±3 mm |
Summenformel | C |
Synonym | Activated charcoal |
Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.