missing translation for 'onlineSavingsMsg'
Learn More
Acetone, Analytical Grade, Penta Chemicals Artikelnummer.: 30020433
Zum Warenkorb hinzufügen
compliance-icons compliance-icons

Acetone, Analytical Grade, Penta Chemicals

Artikelnummer. 30020433
1 L
Click to view available options
Menge:
1 L
2.5 L
5 L
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 30020433

Marke: PENTA CHEMICALS 1006011000

Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Top quality chemicals for laboratories, schools, hospitals, research institutes, and large manufacturing plants.

About us

The company PENTA was established in 1990 under the trade name Ing. Petr Švec - PENTA s.r.o.. From the very beginning, we have been a solely Czech company which is managed by the Švec family, including company founder Petr Švec.

We specialize in the manufacture and distribution of pure, laboratory, pharmaceutical and specialty chemicals, which we market under the trade name Penta. We manufacture and supply a wide range of organic and inorganic chemicals, in large volumes, with a quality ranging from technical, to pure, to A.G. and ACS.

High quality and flexibility

Thanks to the high quality of our products, and our ability to fulfil specific customer requirements, we rank among the leading suppliers of qualified chemicals in the domestic market, and more recently we have also expanded into the global market.

We manufacture the chemicals in our own modern complex, which fulfils the strictest quality and safety criteria. Our large warehouse facilities, including two automated warehouses, enable us to accomodate client requirements. We package the products in accordance with all legislative requirements, and can deliver them to the customer using our own ADR transport on request.

Environmental responsibility

Our priority is to supply the maximum quality of chemicals, reliability, and environmental awareness. Our efforts are evidenced by many awards and certificates, including ISO 9001, ISO 14001 and the Responsible Care certificate for responsible business in the chemical industry.

Chemische Identifikatoren

CAS 67-64-1
Molekulargewicht (g/mol) 58.08

Spezifikation

Chemischer Name oder Material Acetone
Menge 1 L
Verpackung Glass Bottle
Reinheit (%) ≥99.5%
Güte Analytical
compliance-icons compliance-icons
Produktidentifikator
  • Acetone
Signalwort
  • Gefahr
Gefahrenkategorie
  • Schwere Augenschädigung/Augenreizung Kategorie 2
  • Entzündbare Flüssigkeit Kategorie 2
  • Spezifische Zielorgan-Toxizität Kategorie 3
Gefahrenhinweis
  • H225-Flüssigkeit und Dampf leicht entzündbar.
  • H319-Verursacht schwere Augenreizung.
  • H336-Kann Schläfrigkeit und Benommenheit verursachen.
Sicherheitshinweis
  • P210-Von Hitze, heißen Oberflächen, Funken, offenen Flammen und anderen ndquellen fernhalten. Nicht rauchen.
  • P261-Einatmen von Staub/Rauch/Gas/Nebel/Dampf/Aerosol vermei den.
  • P305+P351+P338-BEI KONTAKT MIT DEN AUGEN: Einige Minuten lang behutsam mit Wasser spülen. Vorhandene Kontaktlinsen nach Möglichkeit entfernen. Weiter spülen.
Ergänzende Informationen
  • MIXTURE LIST-Enthält: acetone
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts
Acetone, Analytical Grade, Penta Chemicals >

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt