CAS RN 100-84-5
CAS RN 100-84-5
IUPAC Name:
1-methoxy-3-methylbenzene
Synonyme:
M-methylanisole
Molekulargewicht (g/mol):
122.17
Summenformel:
C8H10O
InChi Key:
OSIGJGFTADMDOB-UHFFFAOYSA-N
SMILES:
COC1=CC=CC(C)=C1
1
–
1
von
1
Ergebnisse
1
3-Methylanisol, 99 %, Thermo Scientific Chemicals
CAS: 100-84-5 Summenformel: C8H10O Molekulargewicht (g/mol): 122.17 MDL-Nummer: MFCD00008395 InChI-Schlüssel: OSIGJGFTADMDOB-UHFFFAOYSA-N Synonym: 3-methylanisole,3-methoxytoluene,m-methylanisole,m-cresol methyl ether,m-methoxytoluene,benzene, 1-methoxy-3-methyl,anisole, m-methyl,methyl m-tolyl ether,m-cresyl methyl ether,methyl m-cresyl ether PubChem CID: 7530 IUPAC-Name: 1-Methoxy-3-methylbenzol SMILES: COC1=CC=CC(C)=C1