missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Z)-4-Hydroxytamoxifen, Tocris Bioscience™
Click to view available options
Menge:
10 mg
50 mg
Beschreibung
(Z)-4-Hydroxytamoxifen is an estrogen receptor antagonist (IC50 = 27 μM). (Z)-4-Hydroxytamoxifen is a Tamoxifen (Cat. No. 0999) metabolite; that exhibits greater potency than the parent compound. (Z)-4-Hydroxytamoxifen Also activates intein-linked inactive Cas9, reducing off-target CRISPR-mediated gene editing; system has ∽25-fold higher specificity than wtCas9. (Z)-4-Hydroxytamoxifen binds to voltage-gated sodium channels, near the intracellular gate and inhibits the sodium current by delaying channel recovery from the inactivated state (IC50 values are 297 nM and 2.1 μM for NavM and human Nav, respectively in HEK cells); this inhibition is independent of estrogen receptor modulation. Chemotherapeutic agent.
Spezifikation
Spezifikation
Chemischer Name oder Material | (Z)-4-Hydroxytamoxifen |
Menge | 50 mg |
Formelmasse | 387.51 |
Reinheit (%) | >98% |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur