Learn More
Brady™ Workstation Print Partner Software
Ensure label consistency in the production line by creating pre-determined label templates with Workstation Print Partner.
Marke: Brady™ 146248
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Brady™ Workstation Print Partner puts you in complete control of label consistency by separating label design from label printing. Print Partner enables you to ensure label consistency by deploying pre-determined label templates. These cannot be changed by operators in the production line. Via Print Partner, locked label templates can be completed with the necessary information.
- Information can be drawn from Excel™ to optimize process
- Requires a minimum of 1 GB RAM and 200 MB free disk space
Spezifikation
Email with Activation Code | |
Bulgarian, Chinese (Tradditional), Chinese (Simplified), Croatian, Czech, Danish, Dutch, English, Estonian, Finnish, French, German, Hungarian, Italian, Japanese, Norwegian, Polish, Portuguese, Romanian, Russian, Slovakian, Slovenian, Spanish, Swedish, Turkish | |
Windows™ 7, Windows™ 8, Windows™ 10 | |
Electrical Labeling, Laboratory Labeling, Wire and Cable Labeling |