missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ WNK1 Recombinant Protein Antigen

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

Marke:  Novus Biologicals™ NBP2-58438PEP

Artikelnummer. 18205803

  • 291.00 CHF / 100 Mikroliter
Verfügbar ab: 23-12-2024
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Beschreibung

Beschreibung

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human WNK1. The WNK1 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Spezifikation

Spezifikation

65125
>80% by SDS-PAGE and Coomassie blue staining
PBS and 1M Urea, pH 7.4.
WNK1
100 μl
E. coli
Human
Store at -20°C. Avoid freeze-thaw cycles.
Antibody Competition
27 kDa
RUO
This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-54281. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote