Learn More
Tocris Bioscience™ VTP 27999 trifluoroacetate
Highly potent and selective renin inhibitor
Marke: Tocris Bioscience™ 5563/10
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Description
VTP 27999 trifluoroacetate is a highly potent and selective renin inhibitor (IC50 = 0.47 nM). Displays >1000-fold selectivity over related and unrelated off-targets. Blocks renin-stimulated ERK1/2 phosphorylation in vascular smooth muscle cells. Exhibits a different mode of action to aliskiren (Cat. No. 5491).Specifications
Methyl N-[2-[(R)-(3-Chlorophenyl)[(3R)-1-[[[(2S)-2-(methylamino)-3-[(3R)-tetrahydro-2H-pyran-3-yl]propyl]amino]carbonyl]-3-piperidinyl]methoxy]ethyl]carbamate trifluoroacetate | |
1013937-63-7 | |
10 mg |