Learn More
KARTELL Viscous Liquid Pump
The Viscous Liquid Pump is a versatile pump that has the capacity of up to 6 gallons per minute.
Marke:  KARTELL 02004004
Artikelnummer. 10090971
  • 235.00 CHF / 1 Stück
Verfügbar ab: 13-02-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
  • Dispensing pump made of PP operated by a unique lever-action stroke.
  • Delivery of approximately 10 ounces (ab. 300 mL) per stroke, with capability of 6 gallons per minute (ab. 22 to 23 litres/min).
  • Will handle the most viscous materials (equivalent to SAE 90 oil).
  • Spigot orientation of 360° allows versatility in dispensing.
  • Complete with shank.
Spezifikation
Spezifikation
Polypropylen
Liquid Pump
6 gal./min.