Learn More
Novus Biologicals™ UTP6 Overexpression Lysate
Marke:  Novus Biologicals™ NBL1-17680
Artikelnummer. 18293040
  • 820.00 CHF / 0.10 Milligramm
Verfügbar ab: 23-12-2024
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
The UTP6 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Spezifikation
Spezifikation
The lysate was created in HEK293T cells, using plasmid ID RC2647 and based on accession number NM_18428. The protein contains a C-terminal DDK tag.
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
Western Blot
RIPA buffer
Homo sapiens UTP6, small subunit (SSU) processome component, homolog (yeast) (UTP6), mRNA.
Native
Sonderangebote
Nur für Forschungszwecke