Learn More
Thermo Scientific™ USA Standard Sieve, 80 Mesh
Brass frame and Stainless-steel mesh sieves
Marke:  Thermo Scientific™
 Lernen Sie weitere Varianten dieses
Artikelnummer. 11329167
  • 143.79 CHF / 1 Stück
Verfügbar ab: 16-12-2024
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
  • Brass frame, stainless steel mesh sieves meet ASTM E11 requirements and all current amendments
  • Sieves are full height
  • Mesh pores: 177μ ( 0.177mm )
Sieve Depths
  • 3 in. diameter sieves are 1 in. deep
  • 8 in. diameter sieves are 2 in. deep
  • 12 in.diameter sieves are 3 in. deep