Learn More
Thermo Scientific™ USA Standard Sieve, 500 Mesh
Brass frame and Stainless-steel mesh sieves
263.00 CHF
Spezifikation
Typ | Sieve |
---|---|
Material | Stainless Steel |
Zertifizierung/Konformität | ASTM E11 |
Beschreibung | 500 Mesh |
Produktcode | Marke | Tiefe (metrisch) | Außendurchmesser (metrisch) | Preis | Menge & Verfügbarkeit | ||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Tiefe (metrisch) | Außendurchmesser (metrisch) | Preis | Menge & Verfügbarkeit | ||||
11369307
|
Thermo Scientific™
040181.ON |
263.00 CHF
1 Stück |
Lager vorrätigLoggen Sie sich ein, um den Lagerbestand zu sehen
|
||||||
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Brass frame, stainless steel mesh sieves meet ASTM E11 requirements and all current amendments
- Sieves are full height
- 3 in. diameter sieves are 1 in. deep
- 8 in. diameter sieves are 2 in. deep
- 12 in.diameter sieves are 3 in. deep
Spezifikation
Sieve | |
Stainless Steel | |
ASTM E11 | |
500 Mesh |
Sicherheit und Handhabung
Recommended Storage : Ambient temperatures
RUO – Research Use Only