Learn More
GE Healthcare Unmounted, Tritium, 19 x 24 cm, Screen Only, Artikelnummer.: 11320372
GE Healthcare Unmounted, Tritium, 19 x 24 cm, Screen Only,
Artikelnummer. 11320372 Alle ansehen Cytiva Produkte
Dimensions: 24L x 19cmW
Click to view available options
:
Dimensions: 24L x 19cmW
Artikelnummer. 11320372
Marke: GE Healthcare 63003549
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.
Alternative Produkte anzeigen
To detect the weak energy of the 3H signal, these screens are constructed without a protective cellulose acetate overlay. The highly sensitive screens eliminate the need for fluorographic reagents. Optimal results are obtained when the 3H signal is on the surface of the sample and available to penetrate the screen.
  • Requires approximately one-tenth the exposure time of traditional autoradiography film.
  • Ten to one hundred times more sensitive than film, depending on isotope and sample type.
  • Wide linear dynamic range (five orders of magnitude) enables quantitation and visualisation of both weak and strong signals in a single exposure.
  • Reusable screens do not require chemicals, darkroom, or other special treatment for use.
  • Each step can be performed at the lab bench under normal lighting conditions and at room temperature.
  • Results are digitised using a storage phosphor imaging system (e.g., Typhoon, Storm, or PhosphorImager) and quantitated using ImageQuant software.
TRUSTED_SUSTAINABILITY
Spezifikation
Menge Je 1
Produkttyp Unmounted Tritium Screen
Berichtigung von Produktinhalten
Name des Produkts
GE Healthcare Unmounted, Tritium, 19 x 24 cm, Screen Only, >