Learn More
Novus Biologicals™ TUBGCP3 Overexpression Lysate
Marke:  Novus Biologicals™ NBL1-17447
Artikelnummer. 18109619
  • 820.00 CHF / 0.10 Milligramm
Verfügbar ab: 20-12-2024
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
The TUBGCP3 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Spezifikation
Spezifikation
The lysate was created in HEK293T cells, using plasmid ID RC27395 and based on accession number NM_6322. The protein contains a C-terminal DDK tag.
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
Western Blot
RIPA buffer
Homo sapiens tubulin, gamma complex associated protein 3 (TUBGCP3), mRNA.
Native
Sonderangebote
Nur für Forschungszwecke