missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSH-Receptor, B-Chain Mouse, Clone: TSHRB/1405, enQuireBio™
Mouse Monoclonal Antibody
234.00 CHF - 484.00 CHF
Spezifikation
Antigen | TSH-Receptor, B-Chain |
---|---|
Klon | TSHRB/1405 |
Konzentration | 200μg/mL |
Anwendungen | Immunohistochemistry |
Klassifikation | Monoclonal |
Spezifikation
TSH-Receptor, B-Chain | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
7253 | |
Recombinant human TSHR, B-Chain protein | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
42kDa |
TSHRB/1405 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
XP_005268094 | |
TSHR | |
IgG2a | |
Protein A/G Purified | |
Cell Surface and Cytoplasmic | |
293T Cells. Thyroid gland |