missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Transglutaminase 1/TGM1 Recombinant Protein Antigen
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Marke: Novus Biologicals™ NBP2-34062PEP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human TGM1. The Transglutaminase 1/TGM1 Recombinant Protein Antigen is derived from E. coli. The Transglutaminase 1/TGM1 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.This is a blocking peptide for NBP2-34062. This is a human recombinant protein expressed in E. coli with a N-terminal His-ABP tag and purified with IMAC chromatography.
Spezifikation
7051 | |
Chromatography | |
0.5mg/mL | |
PBS and 1M Urea, pH 7.4. | |
TGM1 | |
28kDa | |
0.1mL | |
E.Coli | |
MMDGPRSDVGRWGGNPLQPPTTPSPEPEPEPDGRSRRGGGRSFWARCCGCCSCRNAADDDWGPEPSDSRGRGSSSGTRRPGSRGSDSRRPVSRGSGVNAA |
Human | |
>80% | |
Store at -20°C. Avoid freeze-thaw cycles. | |
Blocking/Neutralizing, Control | |
Unmarkiert | |
Transglutaminase 1/TGM1 | |
RUO | |
This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-34062. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml |
Nur für Forschungszwecke