missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transferrin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 17435-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Transferrins (Tf) are an approximately 80kDa blood plasma glycoprotein synthesized by the liver. They are iron binding transport proteins which can bind two Fe(3+) ions in association with the binding of an anion, usually bicarbonate. It is responsible for the transport of iron from sites of absorption and heme degradation to those of storage and utilization. Transferrin is the primary blood iron transport protein and under normal conditions; approximately one-third of total blood transferrin contains bound iron. Measurement of blood transferrin levels can be used as an indicator for blood iron-carrying capacity and abnormalities of iron metabolism such as anaemia, iron overload and haemochromatosis. Serum transferrin may also have a further role in stimulating cell proliferation.Spezifikation
Transferrin | |
Polyclonal | |
Unconjugated | |
TF | |
Beta 1 metal binding globulin, PRO1557, PRO2086, Serotransferrin, Siderophilin, TF, Transferrin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22041, 24825, 7018 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P02787, P12346, Q921I1 | |
TF | |
Transferrin Fusion Protein Ag11504 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |