Learn More
Thermo Scientific™ TopVision Low Melting Point Agarose Promotion
Perform in-gel enzymatic process experiments and nucleic acid recovery after electrophoresis with Thermo Scientific™ TopVision™ Low Melting Point Agarose.
Marke: Thermo Scientific™ R0801
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Top Vision Low Melting Point Agarose is highly purified agarose with a low melting temperature certified by strict quality control test procedures. It is used for in-gel enzymatic process experiments and for nucleic acids recovery from gels using Agarase (#EO0461).
Highlights
- Optimal concentration between 0.7-2% in all typical buffer systems
- GQ (Genetic Quality) certified, which ensures that nucleic acids recovered from preparative gels can be used for downstream applications (enzymatic reactions etc.)
- Low DNA/RNA binding
- Excellent gel transparency
- DNase and RNase free
- Suitable for RNA analysis
Spezifikation
Agarose | |
25 g |