Learn More
TNP1 Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech CL488-17178-100UL
Additional Details : Gewicht : 0.00010kg
Artikelnummer. 17254676
  • 350.75 CHF / 100 Mikroliter
Verfügbar ab: 19-07-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Transition nuclear proteins (TNP1 and TNP2) are the major nuclear proteins that replace somatic histones during spermatogenesis. TNPs are required for normal chromatin condensation and functional sperm development, spermatogenesis was found to be compromised in both Tnp1 and Tnp2 null mice. TNP1, or TP1, localized in nucleus, is a spermatid-specific product of the haploid genome which replaces histone and is itself replaced in the mature sperm by the protamines. Recently, TNP-1 was used as a germ cell marker in condensing spermatids.
Spezifikation
Spezifikation
TNP1
Polyclonal
CoraLite 488
100 μL
7141
Liquid
Immunofluorescence
1000 μg/mL