Learn More
Novus Biologicals™ TMEM186 Overexpression Lysate
Marke:  Novus Biologicals™ NBL1-17049
Artikelnummer. 18293500
  • 820.00 CHF / 0.10 Milligramm
Verfügbar ab: 23-12-2024
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
The TMEM186 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Spezifikation
Spezifikation
The lysate was created in HEK293T cells, using plasmid ID RC24141 and based on accession number NM_15421. The protein contains a C-terminal DDK tag.
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
Western Blot
RIPA buffer
Homo sapiens transmembrane protein 186 (TMEM186), mRNA.
Native
Sonderangebote
Nur für Forschungszwecke