Learn More
Novus Biologicals™ TMEM143 Overexpression Lysate
Marke: Novus Biologicals™ NBL1-17016
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The TMEM143 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.Spezifikation
The lysate was created in HEK293T cells, using plasmid ID RC2449 and based on accession number NM_18273. The protein contains a C-terminal DDK tag. | |
0.1 mg | |
Protein | |
-80°C. Avoid freeze-thaw cycles. |
Western Blot | |
RIPA buffer | |
Homo sapiens transmembrane protein 143 (TMEM143), mRNA. | |
Native |
Nur für Forschungszwecke