missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIM-3 Recombinant Rabbit Monoclonal Antibody (024), Biotin, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen MA552198
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
TIM-3 Recombinant Monoclonal antibody specifically detects TIM-3 in Human samples. It is validated for ELISASpezifikation
TIM-3 | |
Recombinant Monoclonal | |
100 μg/mL | |
PBS with no preservative | |
Q8TDQ0 | |
Rabbit | |
Protein A | |
RUO | |
84868 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA | |
024 | |
Biotin | |
Havcr2 | |
CD366; FLJ14428; Havcr2; HAVcr-2; Hepatitis A virus cellular receptor 2; hepatitis A virus cellular receptor 2 homolog; kidney injury molecule-3; KIM-3; sCD366; soluble CD366; soluble TIM 3; T cell immunoglobulin mucin 3; T cell immunoglobulin mucin-3; T-cell immunoglobulin and mucin domain containing 3; T-cell immunoglobulin and mucin domain-containing protein 3; T-cell immunoglobulin mucin family member 3; T-cell immunoglobulin mucin receptor 3; T-cell membrane protein 3; Tim3; TIM-3; TIMD3; TIMD-3 | |
Recombinant Human TIMD3 protein (Ser 22-Arg200) | |
20 μg | |
Primary | |
Human | |
Antibody | |
IgG |