missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIM-1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA598302
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CD365 also known as T-cell immunoglobulin and mucin domain 1 (Tim-1) is a type 1 surface glycoprotein that consists of extracellular N-terminal immunoglobulin V-like domain and a mucin domain, and a short cytoplasmic tail. The endogenous ligand for CD365 is Tim-4. In addition, CD365 is known to be a cellular receptor or co-receptor for a number of viruses including hepatitis A and C viruses, Ebola virus, Marburg virus, Dengue virus, Zika virus, West Nile virus, and Japanese encephalitis virus. The direct binding of the viruses by CD365 occurs via phosphatildyl-serine exposed on viral envelopes. Polymorphisms within human CD365 gene (HAVCR1) are associated with susceptibility to infection and the disease severity. CD365 has also been implicated in the T cell co-stimulation. CD365 is primarily expressed in epithelial cells. It is also highly expressed in A549, 769-P and Caco-2 and cell lines. CD365 exists also as a soluble protein generated by metalloproteinase mediated proteolytic cleavage of the surface protein.Spezifikation
TIM-1 | |
Polyclonal | |
Unconjugated | |
Havcr1 | |
AI503787, CD365, HAVCR, HAVCR1, HAVcr-1, Hepatitis A virus cellular receptor 1, hepatitis A virus cellular receptor 1 homolog, Kidney injury molecule 1, Kim1, KIM-1, T cell immunoglobin domain and mucin domain protein 1, t cell immunoglobulin and mucin domain-containing protein 1, T cell membrane protein 1, T-cell immunoglobulin and mucin domain containing 1, T-cell immunoglobulin and mucin domain-containing protein 1, T-cell immunoglobulin mucin family member 1, T-cell immunoglobulin mucin receptor 1, T-cell membrane protein 1, TIM, TIM1, TIM-1, TIMD1, TIMD-1 | |
Rabbit | |
Protein G | |
RUO | |
171283, 26762 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1.0 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300, pH 7.4 | |
Q5QNS5, Q96D42 | |
Havcr1 | |
Recombinant Human Hepatitis A virus cellular receptor 1 protein (21-290AA) | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |