missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIM-1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA186790
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
In Western blot, this antibody detects a band at ∽49kDa in human uterus tissue lysate. Rabbit anti TIM-1 antibody detects T cell immunoglobulin mucin-1 (TIM-1), which was first identified as the receptor used by hepatitis A for viral entry, it is also known as the hepatitis A virus cellular receptor- HAVCR (Curtiss and Colgan 2007), TIM -1 also acts as a receptor for other viruses, including Dengue, Ebola and Marburg (Kondratowicz et al. 2011).
CD365 also known as T-cell immunoglobulin and mucin domain 1 (Tim-1) is a type 1 surface glycoprotein that consists of extracellular N-terminal immunoglobulin V-like domain and a mucin domain, and a short cytoplasmic tail. The endogenous ligand for CD365 is Tim-4. In addition, CD365 is known to be a cellular receptor or co-receptor for a number of viruses including hepatitis A and C viruses, Ebola virus, Marburg virus, Dengue virus, Zika virus, West Nile virus, and Japanese encephalitis virus. The direct binding of the viruses by CD365 occurs via phosphatildyl-serine exposed on viral envelopes. Polymorphisms within human CD365 gene (HAVCR1) are associated with susceptibility to infection and the disease severity. CD365 has also been implicated in the T cell co-stimulation. CD365 is primarily expressed in epithelial cells. It is also highly expressed in A549, 769-P and Caco-2 and cell lines. CD365 exists also as a soluble protein generated by metalloproteinase mediated proteolytic cleavage of the surface protein.Spezifikation
TIM-1 | |
Polyclonal | |
Unconjugated | |
Havcr1 | |
AI503787; CD365; HAVCR; HAVCR1; HAVcr-1; Hepatitis A virus cellular receptor 1; hepatitis A virus cellular receptor 1 homolog; Kidney injury molecule 1; Kim1; KIM-1; T cell immunoglobin domain and mucin domain protein 1; t cell immunoglobulin and mucin domain-containing protein 1; T cell membrane protein 1; T-cell immunoglobulin and mucin domain containing 1; T-cell immunoglobulin and mucin domain-containing protein 1; T-cell immunoglobulin mucin family member 1; T-cell immunoglobulin mucin receptor 1; T-cell membrane protein 1; TIM; TIM1; TIM-1; TIMD1; TIMD-1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
171283, 26762, 286934 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O54947, Q5QNS5, Q96D42 | |
Havcr1 | |
16 amino acid peptide sequence near the center of human TIM-1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |