missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TIM-1/KIM-1/HAVCR Antibody Pair [HRP]
Rabbit Monoclonal Antibody Pair
531.00 CHF - 1285.00 CHF
Spezifikation
Assay-Bereich | 125 pg/mL |
---|---|
Assay-Empfindlichkeit: | 125-8000 pg/mL |
Assay | Sandwich ELISA |
Konjugat | HRP |
Immunassay-Kit-Format | Sandwich-ELISA-Kit |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18286426
|
Novus Biologicals™
NBP2-79614-15PLATES |
15 Plates |
1285.00 CHF
15 Stück |
Verfügbar ab: 03-01-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
18210137
|
Novus Biologicals™
NBP2-79614-5PLATES |
5 Platten |
531.00 CHF
5 Stück |
Verfügbar ab: 03-01-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Canine TIM-1/KIM-1/HAVCR.Spezifikation
125 pg/mL | |
Sandwich ELISA | |
Sandwich-ELISA-Kit | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Canine TIM-1/KIM-1/HAVCR. | |
Sandwich-ELISA | |
26762 | |
Rabbit | |
RUO | |
TIM-1/KIM-1/HAVCR |
125-8000 pg/mL | |
HRP | |
Antibody Pairs | |
Hund | |
HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1 | |
HAVCR1 | |
Storage is content dependent. | |
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 1 μg/mL in detection antibody dilution buffer before use. 3. Standard: After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 8 ng/mL is recommended. |